Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 58041EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0226225011EA
Encompass
Three centrifuges in one a high-capacity general-purpose centrifuge for cell harvesting a high-speed centrifuge for separating cell lysates and a microcentrifuge for DNA precipitations. It allows for molecularapplications in tubes up to 250 mL and offers additional wing-bucket and fixed-angle rotors as well as deepwell plate capacity for increased versatility.
Catalog No. 50-290-8975
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.