Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Combitips Advan
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0030089804100EA
Encompass
The Eppendorf Combitips have been completely redesigned and optimized to meet the increased requirements of a modern laboratory to a greater extent than ever before. The result is the Combitips advanced.The Combitips advanced have replaced Combitips plus.
Catalog No. 50-290-1133
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.