Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Combitips advan
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z763055100EA
Encompass
The Combitips advanced are optimized for all Repeater models to create the perfect connectionThe new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality Sterile PCR Clean and Eppendorf Biopur purity grades you will always find the perfect Combitip for your application.Sterile grade makes these products applicable for all applications where the absence of living organisms like microorganisms could interfere the positive outcome of an experiment. They are also Pyrogen free that ensures absolute absence of endotoxines which is mandatory for many applications in cell culture pharmacology and medical research.
Catalog No. 50-290-5996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.