Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Deepwell Plate
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP95103358820EA
Encompass
Eppendorf(R) Deepwell Plates 96 and 384 are high-performance plates for all manual and automated applications - from sample storage at -86 AºC to DNA denaturation at 100 AºC. Manufactured from pure polypropylene (PP) these plates offer innovative features More dependability. More efficiency. More stability. The plates are available with a white yellow green or blue border. for easy identification of wells a high-contrast alphanumeric label is laser-engraved in the border. This OptiTrack(R) matrix allows you to find your sample up to 30 % more quickly while reducing pipetting errors. With all of these sophisticated features these plates set new standards in your laboratory.
Catalog No. 50-290-1645
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.