Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR DNA LoBind Tube
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0030108310200EA
Encompass
Eppendorf Tubes are the perfect option for working with medium-sized sample volumesUL LIMaximum safety and stability for centrifugation up to 25 000 x g/LILIConical bottoms/LILIEasier processing of samples up to 5.0 mL/LILIHigher yields in DNA isolation prep of mastermixes and buffers and safe cell and tissue lysis/LILITransparent polypropylene free of plasticizers biocides or mold release agents/LILIHinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86C to 80C/LI/UL
Catalog No. 50-290-1530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.