Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR epT.I.P.S. Biop
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z640174100EA
Encompass
Biopur guaranteed sterile pyrogen-free RNase-free DNA-free and ATP-free for applications in pharmaceutical and food industries molecular biology and cell technology.Batch specific certificates available on the internet at http//www.eppendorf.com
Catalog No. 50-290-0838
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.