Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR epT.I.P.S.R G R
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP00300760441PK
Encompass
Designed for a cone distance of 4.5 mmepT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16 and 24 channel pipettes for utmost safety and reliabilityInnovative SOfTattach technology allows for reliable tip fit and tightnessfill a whole 384 well plate without loosening pipette tipsPerfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transferThe tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384 well platesTip attachment forces reduced by 40% per cone compared to 8 and 12 channel pipettesTip ejection forces reduced by SOfTattach for ergonomic working conditionsAvailable as reload and setAvailable in purity grades Eppendorf Quality and PCR clean
Catalog No. 50-290-4792
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.