Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR heat sealing fo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0030127854100EA
Encompass
Eppendorf(R) heat sealing foil is used for continuous locking with the best evaporation protection during PCR. It is made of aluminum can be pierced and protects light-sensitive samples.
Catalog No. 50-290-1808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.