Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR MastercyclerR X1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP63010000121EA
Encompass
The Eppendorf Mastercycler X50 Thermocycler combines speed flexibility and PCR optimization for research PCR applications and standardization for routine PCR applications such as food testing. A highly intuitive touch screen puts all these benefits at your fingertips.Speed and enhanced PCR optimization functions like the 2D-Gradient make the Eppendorf Mastercycler X50 Thermocycler the ideal PCR cycler for advanced research in molecular biology. The excellent block temperature regulation gives rise to the next stage of PCR reproducibility whereas the adaptable user management and profound documentation capabilities give peace of mind to laboratories working to specific or strict standards.A highly intuitive touch display low noise levels low power consumption and the versatile flexlid concept combine to make a powerful PCR cycler. Up to 10 PCR cyclers can be combined - ideal for high throughput applications or labs with a high number of users running different assays. Should you f
Catalog No. 50-290-8828
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.