Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR ReferenceR 2 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP49240009083EA
Encompass
Extraordinary precision and accuracy - EppendorfA´s premium pipetteBesides improved ergonomics the design focus lays on providing the most accurate results possible optimum user safety reliability and robustness for a long service life. The innovative single button operation is not only fast and easy but it also allows active aerosol1) reduction and thus protects the user the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility Eppendorfs Reference 2 is an ideal instrument for these liquid handling applications.Single-Button OperationLower Operating forces for aspiration/dispensing
Catalog No. 50-290-9103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.