Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR RepeaterTM E3/E1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP49870001181EA
Encompass
The MultipetteAE3/E3x in combination with the Combitips(R) advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications accommodating more than 5 000Adifferent dispensing volumes between 1AmuL to 50AmL.
Catalog No. 50-290-8691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.