Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR RepeaterTM E3/E1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP49870001341EA
Encompass
These electronic dispensers combine easy handling during dispensing with a broad range of features The Repeater E3x offers additional high-end applications such as titration and aspirating of supernatants or sequential dilutions. In combination with the Combitips advanced it is an essential lab tool for all kind of liquid handling.
Catalog No. 50-290-8503
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.