Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR RepeaterTM M4 s1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP49820003221EA
Encompass
With the RepeaterAM4/Combitip System volumes are dispensed using the positive displacement principle. The liquid is directly dispensed without an air cushion ensuring the correct volume is always dispensed regardless of the density viscosity and volatility of the liquid. The RepeaterAM4 offers a unique volume range of 1AmuLA-A10AmL and is the only mechanical dispenser that dispenses the aspirated liquid up to 100Atimes without refilling the tip. The innovative step counter supports error-free working even after an interruption or distraction during the dispensing procedure. A fully emptied Combitip can be easily ejected with one hand using the operation lever without directly touching the tip.
Catalog No. 50-290-9122
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.