Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR ResearchR plus
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP22313000064EA
Encompass
The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy - combined with ultimate ergonomics and increased flexibility. The pipettes features a spring loaded tip cone a secondary calibration option an improved volume display and is fully autoclavable.The Eppendorf Research plus has been developed within the strict criteria of the Eppendorf PhysioCare Concept(R). PhysioCare Concept(R) stands for ultimate durability intuitive operation and above all for perfect balance and minimal user exertion.Eppendorf Research plus pipette provides flexibility. Calibrate your Research plus to your needs autoclave the entire pipette or only the lower part. Choose among single channel multi channel and fix-volume pipettes in different sizes.Volume adjustment Only a few turns to reach your desired volumeControl button and Ejector Very low operating force color indicates pipette volume positioned for perfect ergonomicsVolume display 4 digits
Catalog No. 50-290-8941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.