Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Safe-Lock Micro
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP022600044100EA
Encompass
Patented Safe-Lock hinged lid that eliminates any danger from leaks or from opening accidentallyCan be autoclaved when open (121C 20 minutes)Has volume graduations and etched marking surfacesIndividually sealed Biopur microcentrifuge tube is sterile and free of pyrogens RNase DNA and ATP
Catalog No. 50-290-1583
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.