Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR ThermoMixerR C1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP53820000231EA
Encompass
You need everything? The Eppendorf ThermoMixer C combines excellent mixing performance with excellent temperature control to guarantee complete dependable and reproducible test results. Improve your assay results by mixing and incubating samples at the same time. The Eppendorf ThermoMixer C convinces users with its quick and easy-to-exchange SmartBlocks for all common lab vessels new and improved program functions and high level of stability for every mixing frequency. There is almost no limit to the variety of applications waiting for you2DMix-Control The unique 2DMix-Control technology mixes liquids in a controlled circular movement. The ThermoMixer C provides enough mixing power to handle the most challenging mixing tasks. Most samples are efficiently mixed within less than a minute independent of vessel geometry or sample properties.A wide varity of Eppendorf SmartBlocks are available for the Eppendorf ThermoMixer C. These thermobocks offer the utmost flexibility for use of all
Catalog No. 50-290-8948
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.