Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Tubes 5 mL Scre
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0030122364400EA
Encompass
Original Eppendorf(R) PCR tubes are manufactured according to the highest Eppendorf quality standards.Made of exceptionally high-quality transparent polypropylene free of plasticizers biocides or mold release agents for reliable test resultsThese thin-walled polypropylene tubes ensure efficient and homogeneous heat transfer. They are easy to open but provide tight sealing to prevent evaporation during PCR. Efficient heat transfer to the sample is ensured thanks to their thin even wall thickness and smooth wall surface.Centrifugation safety Maximum safety and stability for centrifugation up to 25 000 x g. fast and efficient protocols.features screw cap.
Catalog No. 50-290-4067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.