Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR XplorerR 8 chan1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP48610001631EA
Encompass
Multi-functional rocker with up is up and down is down functionalitySelection dial for quick and easy selection of functionsSelf-explanatory color display without submenus and with ergonomic angle for optimal readability in every positionSpring-loaded tip cone for all pipettes up to 1 000 muL for minimal tip attachment forces helps to reduce stressPowerful rechargeable battery for up to 8 hours work without the need for rechargingSeparate power socket Work can continue while chargingAdjustment to external conditions (e.g. geographic height liquids with high density)Lower part of pipette can be autoclaved to ensure decontaminationSpring-loaded tip cone (not for 1 200?muL multi-channel)Xplorer plus only Individual programming fixed volume settings password protection on/off switch additional applications display of service intervals
Catalog No. 50-290-8943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.