Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EPPTB
GREENER_CHOICE

Catalog No. p-7110774
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-185-0 50 mg
45-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-185-0 Supplier Tocris Bioscience™ Supplier No. 4518/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Trace amine 1 (TA1) receptor antagonist/inverse agonist

EPPTB is a trace amine 1 (TA1) receptor antagonist/inverse agonist; exhibits a higher potency at the mouse TA1 receptor than the rat and human TA1 receptors (IC50 values are 27.5, 4539 and 7487 nM, respectively). Blocks the TA1 receptor-mediated activation of an inwardly rectifying K+ current, which increases the firing frequency of dopamine (DA) neurons in the ventral tegmental area. Increases DA potency at the D2 receptor. Also displays inverse agonism, reducing basal cAMP levels in vitro (IC50 = 19 nM).

Specifications

Chemical Name or Material N-(3-Ethoxyphenyl)-4-(1-pyrrolidinyl)-3-(trifluoromethyl)benzamide
CAS 1110781-88-8
Quantity 50 mg
Target Trace Amine 1 (TA1) Receptor Antagonists
Molecular Formula C20H21F3N2O2
Formula Weight 378.39
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.