Learn More
Medchemexpress LLC HY-128849 500mg Medchemexpress, Eprodisate CAS:21668-77-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128849500MG
Encompass_Preferred
Medchemexpress, HY-128849 500mg Eprodisate CAS:21668-77-9 Eprodisate is a new compound designed to interfere with interactions between amyloidogenic proteins and glycosaminoglycans and thereby inhibit polymerization of amyloid fibrils and deposition of the fibrils in tissues. Eprodisate slow the progression of AA amyloidosis-related renal disease and has possible applicability to other types of amyloidosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.