Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103090 Supplier Thermo Scientific Supplier No. PA5103090
Only null left
Add to Cart
Add to Cart

EPS8 Polyclonal Antibody

EPS8 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPS8
Gene Accession No. Q12929
Gene Alias AI042819; AW261790; AW545405; DFNB102; epidermal growth factor receptor kinase substrate 8; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; Eps8r2; EPS8-related protein 2
Gene Symbols EPS8
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPS8(Accession Q12929), corresponding to amino acid residues D555-T605.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2059
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.