Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus Early Antigen Monoclonal Antibody (1108-1)
GREENER_CHOICE

Catalog No. 01672057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-057 Supplier Invitrogen™ Supplier No. MA135751
  • $594.00
    $487.70 / Each of 1

    Save $106.30

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1108-1 Recognizes a 55-50 kDa polypeptide associated with the early antigen of Epstein-Barr virus (EBV). p55 Has beenshown to be a phosphoprotein and p55-50 has strong DNA-binding activity preferentially to single-stranded DNA. Epstein-Barr virus is the causitive agent of infectious mononucleosis and is associated with two human neoplasms, Burkitt's lymphoma and nasopharyngeal carcinoma. Several EBV-related antigens associated with early or late functions of the viral genome have been identified. The early antigen may be virally or chemically induced in EBV infected cells and is the first detectable marker of EBV infection in human cells.

Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus Early Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 1108-1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias EBV; EBV EA; EBV early
Host Species Mouse
Immunogen EBV early antigens.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.