Learn More
ACROBiosystems Epstein-Barr virus Glycoprotein H & Glycoprotein L Heterodimer, Twin-Strep&His Tag (GHL-E5283) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Leu 682 (gH) & Asn 23 - Gly 137 (gL) (Accession # P03231 (gH) & P03212 (gL)).
Supplier:  ACROBiosystems GHLE5283100UG
Encompass_Preferred
Epstein-Bar Virus (EBV), belonging to gamma herpesvirus, is one of the most common human viruses which widely spread all over the world. EBV is the primary cause of infectious mononucleosis (IM) and is found to be related to several types of cancers. Although gp350 is the most abundant glycoprotein on viruses’ surfaces, the research on using gp350 as vaccine antigen progresses slowly making glycoprotein H and glycoprotein L heterodimer (gH/gL) be considered as a candidate for vaccine development. gH/gL plays an important role in assisting the fusion process with the binding of Ephrin type-A receptor 2 (EphA2).
Catalog No. 50-223-1205
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.