Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPSTI1 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145424
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145424 20 μL
FEPA5145423 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145424 Supplier Invitrogen™ Supplier No. PA5145424
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPSTI1 was up-regulated in breast carcinomas. The exact function of EPSTI1 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPSTI1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene EPSTI1
Gene Accession No. Q96J88
Gene Alias 2310046K10Rik; 5033415K03Rik; BRESI1; epithelial stromal interaction 1; epithelial stromal interaction 1 (breast); epithelial-stromal interaction protein 1; EPSTI1; RGD1563207; Unknown (protein for IMAGE:7987136)
Gene Symbols EPSTI1
Host Species Rabbit
Immunogen Recombinant Human EPSTI1 protein, Asn73-Glu280 (Accession #Q96J88).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100625050, 94240
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.