Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ epT.I.P.S.™ 384 Reloads, Eppendorf Quality™,

Catalog No. 05413609 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
20 to 0.1 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-413-609 20 to 0.1 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-413-609 Supplier Eppendorf™ Supplier No. 0030076028

missing translation for 'validMainframeMsg'

epT.I.P.S. 384 micropipette tips with innovative SOFTattach technology are optimally coordinated to Eppendorf 16 and 24-channel pipettes Research plus and Xplorer plus. Process 384-well plates manually with highest level of tip tightness and coaxiality but extraordinary low operating forces.

  • Designed for a cone distance of 4.5 mm
  • epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16 and 24-channel pipettes for utmost safety and reliability
  • Innovative SOFTattach technology allows for reliable tip fit and tightness
  • Fill a whole 384-well plate without loosening pipette tips
  • Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Tip attachment forces reduced by 40 % per cone compared to 8 and 12-channel pipettes
  • Tip ejection forces reduced by SOFTattach for ergonomic working conditions
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 to 0.1 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model epT.I.P.S.™ 384 Set
Length (English) 1.7 in.
Length (Metric) 42 mm
Clean Claims Biopur™, Eppendorf Quality™, PCR clean, Sterile
Storage Requirements Room temperature
Format Hinged Rack
Tip Style Universal
Packaging 5 Trays x 384 Tips
Quantity 1920 Tips
Product Type Micropipette Tips
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less

The pipette tips of the :Productname: product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned :Productname: are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.