Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ epT.I.P.S.™ Box 2.0

Catalog No. 01675128 Shop All Eppendorf North America Products
Change view
Click to view available options
Size:
10 to 300 μL
1000 to 1250 μL
1250 μL to 5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-675-128 1000 to 1250 μL
01-675-150 10 to 300 μL
01-675-156 1250 μL to 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-675-128 Supplier Eppendorf™ Supplier No. 0030076257
Add to Cart
Add to Cart

Eppendorf epT.I.P.S.™ Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid™ technology, enhanced stackability, and superior ergonomics for safe, organized pipetting.

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White, Blue
Holds Pipette Tips
Material Polypropylene
Positions Multiple
Shape Square
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) epT.I.P.S reloads
Packaging 1 Empty Reusable Box
Quantity 1 Ea.
Size 1000 to 1250 μL
Includes Box Only
Product Type Empty Tip Box
Array None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.