Learn More
Apexbio Technology LLC EPZ5676,10mg CAS# 1380288-87-8
SDP
Supplier:  Apexbio Technology LLC A416610
Encompass_Preferred
EPZ5676 is a potent inhibitor of DOT1L histone methyltransferase, according to X-ray crystallographic analysis, that occupies the S-adenosyl methionine (SAM) binding pocket of DOT1L and induces conformational changes in DOT1L resulting in the opening of a hydrophobic pocket beyond the amino acid portion of SAM. EPZ5676 selectively inhibits DOTIL with a value of 50% inhibition concentration IC50 of 0.8 nM, which is 37000-fold greater in selectivity than other methyltransferases, including CARM1, EHMT1/2, EZH1/2, PRMT1/2/5/6/8, SETD7, SMYD2/3, and WHSC1/1L1. EPZ5676 has been investigated for the treatment of MLL-rearranged leukemia in multiple studies where results have shown that EPZ5676 inhibits H3K79 methylation and the expression of MLL-fusion target gene and potently kills acute leukemia cell lines bearing MLL translocation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0011
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.