Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Equal Mass Metal Cylinder Activity Set
SCI_ED

Catalog No. S24865 Shop All Eisco Products
Change view
Click to view available options
Quantity:
Set of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24865 Set of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24865 Supplier Eisco™ Supplier No. PH0112FCBLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For beginner students to understand concepts of density.

  • With five cylinders, each made of aluminum, copper, tin, zinc, brass
  • Density of metals range from 2.7g/cm to 8.8g/cm
  • Instruction manual provides background on density and specific gravity
  • With three inquiry-based student activities with capture sheets
  • Lead-free

Specifications

Quantity Set of 5
For Use With (Application) Density experiments
Diameter (English) 0.5 in.
Product Type Equal Mass Metal Cylinder Activity Set
Includes Set of five cylinders: zinc, copper, aluminum, tin and brass
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.