Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Equal Mass Metal Cylinders, Set Of 5
SDPSCI_ED

Catalog No. S00657 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00657 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00657 Supplier United Scientific™ Supplier No. SGMC05
Add to Cart
Add to Cart
This item is not returnable. View return policy

Equal Mass Metal Cylinders Set is a highly diverse set of samples with the same mass.

  • Each specimen in this set of five diverse materials cylinders has identical mass and radius.
  • Materials included are aluminum, zinc, tin, steel and copper.
  • Includes a storage box and activity guide.

Specifications

Grade 6 to 12
Format Set
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.