Learn More
United Scientific Supplies Equal Volume Squares
SDPSCI_ED
Catalog No. S07039 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Aluminum
Aluminum, Copper, Steel, Brass
Product Type:
Equal Volume Metal Squares Set
Metal Square Block
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
S07039 Aluminum, Copper, Steel, Brass Equal Volume Metal Squares Set
S07027 Aluminum Metal Square Block
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S07039 Supplier United Scientific Supplies Supplier No. SGVS04
Add to Cart
Add to Cart
Used for specific gravity experiments.
  • Available as individual squares
  • Made of copper, brass, steel and aluminum
Specifications
For Use With (Application) Measure the specific gravity
Product Type Equal Volume Metal Squares Set
Includes Squares made of Copper, Aluminum, Brass and Steel, set up box
Material Aluminum, Copper, Steel, Brass