Learn More
Kingfisher Biotech Inc Equine Annexin V Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1797E025
Encompass_Preferred
The Equine Annexin V yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine Annexin V applications are for cell culture, ELISA standard, and Western Blot Control. Equine Annexin V yeast-derived recombinant protein can be purchased in multiple sizes. Equine Annexin V Specifications: (Molecular Weight: 36.0 kDa) (Amino Acid Sequence: MAQVLKGTVT DFPGFDERAD AEVLRKAMKG LGTDEESILT LLTSRSNAQR QEIAVAFKTL FGRDLLDDLK SELTGKFEKL IVALMKPSRL YDAYELKHAL KGAGTNEKVL TEIIASRTPA ELRAIKQVYE EEYGSNLEDD VVADTSGFYQ RMLVVLLQAN RDPDAGIDEA QVEQDAQTLF QAGELKWGTD EEKFITIFGT RSVSHLRRVF DKYMTISGFQ IEETIDRETS GNLEQLLLAV VKSIRSIPAY LAETLYYAMK GAGTDDHTLI RVVVSRSEID LFNIRKEFRK NFATSLYSMI KSDTSGDYKK ALLLLCGGED D (321)) (Gene ID: 100072241). For research use only. Made in the USA
Catalog No. 50-253-2757
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.