Learn More
Alpco Ltd Equine C-reactive Protein ELISA (Equine CRP ELISA)
SDP
Supplier:  Alpco Ltd 41CRPEQE01
Encompass_Preferred
The Equine C-reactive Protein ELISA (Equine CRP ELISA) is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring CRP in horse biological samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.