Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Equine Donor Serum

Catalog No. ICN2921154 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
Color:
Golden Brown
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
ICN2921154 500 mL Golden Brown
ICN29211Z0 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN2921154 Supplier MP Biomedicals™ Supplier No. 092921154

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Horse
Form Liquid
Product Type Horse Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Diagnostic assays
Hemoglobin Concentration ≤35mg/dL
Mycoplasma Detected No
Osmolality 273 to 301 mOsm/kg
pH 7.1 to 8
Protein Content 5.6 to 9.1 g/dL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.