Learn More
Raybiotech Inc Equine IL-10 ELISA Kit
Supplier: Raybiotech Inc ELEIL101
For cell culture supernatants, plasma, and serum samples. Sensitivity: 90 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Equine IL-10.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.