Learn More
Kingfisher Biotech Inc Equine IL-28B Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1279E005
Encompass
The Equine IL-28B yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine IL-28B applications are for cell culture, ELISA standard, and Western Blot Control. Equine IL-28B yeast-derived recombinant protein can be purchased in multiple sizes. Equine IL-28B Specifications: (Molecular Weight: 19.0 kDa) (Amino Acid Sequence: GPVPTSKPAT TQRSCDIGRF KSLSPSELEA FKKAKDALEN SLENWTCRSP VFPTTWDLRQ LQVWERPVAL EAELALTLKI LAMVADSSLA DILDRPLHTL RHIHSELQAC VPARPTAGPG PRGRLHQWLR QLHEVQKKES WGCLEASVTF NLFRLLTRDL NCVASGDLCL (170)) (Gene ID: 100146825). For research use only. Made in the USA
Catalog No. 50-253-3449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.