Learn More
Kingfisher Biotech Inc Equine IL-36RA Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2002E500
Encompass_Preferred
The Equine IL-36 Receptor Antagonist yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine IL-36 Receptor Antagonist applications are for cell culture, ELISA standard, and Western Blot Control. Equine IL-36 Receptor Antagonist yeast-derived recombinant protein can be purchased in multiple sizes. Equine IL-36 Receptor Antagonist Specifications: (Molecular Weight: 17.0 kDa) (Amino Acid Sequence: VLSGALCFRM KDAALKVLYL HDNQLLAGGL HTGKVMKGEE ISVVPNRFLD AKLSPVILGV QGGSQCLSCG TEQEPTLKLE PVNIMELYLG AKEYKSFTFY RRDTGLTSSF ESAAYPGWFL CTVPEADQPL RLTQLPEDAS GDDPITDFYF QQCD (154)) (Gene ID: 100065154). For research use only. Made in the USA
Catalog No. 50-253-2535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.