Learn More
Kingfisher Biotech Inc Equine LIF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2117E005
Encompass
The Equine Leukemia Inhibitory Factor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine Leukemia Inhibitory Factor applications are for cell culture, ELISA standard, and Western Blot Control. Equine Leukemia Inhibitory Factor yeast-derived recombinant protein can be purchased in multiple sizes. Equine Leukemia Inhibitory Factor Specifications: (Molecular Weight: 21.3 kDa) (Amino Acid Sequence: SPLPITPDNA TCATRHPCHS NLMDQIRNQL VQLNSSANAL FILYYTAQGE PFPSNLDKLC RPDVTDFPPF HANGKEKARL VELYRIIAYL GASLGNITRD QKVLNPNALS LHSKLNATSD TMRGLLSNVL CHLCSKYHVA NVDVAYGPDT SNKDVFQKKK LGCQLLGKYK QVIAVVAQAF QTGDLEAYSD PRS (193)) (Gene ID: 100629131). For research use only. Made in the USA
Catalog No. 50-253-2408
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.