Learn More
Kingfisher Biotech Inc Equine TNF alpha Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0930E025
Encompass_Preferred
The Equine TNF alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine TNF alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Equine TNF alpha yeast-derived recombinant protein can be purchased in multiple sizes. Equine TNF alpha Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: LRSSSRTPSD KPVAHVVANP QAEGQLQWLS GRANALLANG VKLTDNQLVV PLDGLYLIYS QVLFKGQGCP STHVLLTHTI SRLAVSYPSK VNLLSAIKSP CHTESPEQAE AKPWYEPIYL GGVFQLEKGD QLSAEINQPN YLDFAESGQV YFGIIAL (157)) (Gene ID: 100033834). For research use only. Made in the USA
Catalog No. 50-241-3234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.