Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Equol, Thermo Scientific Chemicals

Catalog No. p-7042111
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61383MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61383MC Supplier Thermo Scientific Chemicals Supplier No. J61383MC
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Urinary metabolite of daidzein, produced by humans and horses

(±)-Equol exhibits EC50 values of 200 and 74 nM for human ERα and ERβ, respectively and induces breast cancer cell proliferation in vitro at concentrations as low as 100 nM. It is a weak estrogenic agonist, and competitive inhibitor of 17β-estradiol at the estradiol receptor. It inhibits 12-O-tetradecanoylphorbol 13-acetate (TPA)-induced neoplastic cell transformation by targeting the MEK/ERK/p90RSK/activator protein-1 signalling pathway. It functions as a DHT blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-Equol exhibits EC50 values of 200 and 74 nM for human ERα and ERβ, respectively and induces breast cancer cell proliferation in vitro at concentrations as low as 100 nM. It is a weak estrogenic agonist, and competitive inhibitor of 17β-estradiol at the estradiol receptor. It inhibits 12-O-tetradecanoylphorbol 13-acetate (TPA)-induced neoplastic cell transformation by targeting the MEK/ERK/p90RSK/activator protein-1 signalling pathway. It functions as a DHT blocker.

Solubility
Soluble in DMSO and methanol or 100% ethanol. Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94105-90-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00016662
InChI Key ADFCQWZHKCXPAJ-UHFFFAOYNA-N
Synonym 3-4-hydroxyphenyl chroman-7-ol, equol, +/--equol, r,s-equol, 7,4'-homoisoflavane, 4',7-dihydroxyisoflavane, rs-equol, 3-4-hydroxyphenyl-7-chromanol, 2h-1-benzopyran-7-ol, 3,4-dihydro-3-4-hydroxyphenyl, 4',7-isoflavandiol
PubChem CID 382975
IUPAC Name 3-(4-hydroxyphenyl)-3,4-dihydro-2H-chromen-7-ol
SMILES OC1=CC=C(C=C1)C1COC2=C(C1)C=CC(O)=C2

Specifications

Melting Point 172°C to 176°C
Color White
Quantity 100 mg
Beilstein 87752
Sensitivity Hygroscopic
Merck Index 14,3644
Solubility Information Soluble in DMSO and methanol or 100% ethanol. Insoluble in water.
Formula Weight 242.27
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Equol
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.