Learn More
TARGETMOL CHEMICALS INC ER 50891 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22767
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ER-50891 is a potent retinoic acid receptor alpha (RAR(alpha)) antagonist. er-50891 reduces the inhibitory effect of allosteric retinoic acid and restores osteoblast differentiation induced by bone morphogenetic protein 2. purity: 99%
Catalog No. 50-258-0925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.