Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
E.R. Advanced Ceramics Jars for Jar Mills
Manufacturer:  E.R. Advanced Ceramics J7748E
 View more versions of this product
Catalog No. 08-382B
Description
Description
For 08-381 Series Jar Mills.
  • Porcelain fortified with alumina
  • Strong and chemical resistant
  • Extra-wide mouth
  • Locking lid with bar and hand wheel
  • Neoprene gasket
  • Precision-ground cover and gasket seal
Specifications
Specifications
37.19 oz.
37.19 oz., 1100mL
5.75 in.
14.6cm
17.14cm
Fortified Porcelain, Alumina
2.26kg
1100mL
2.75 in.
6.98cm
6.75 in.
Locking lid with bar, hand wheel, Neoprene gasket, precision ground cover, gasket seal
5 lb.
08-381 Series jar mill