Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ER Associated Degradation Antibody Pack
SDP

Catalog No. NBP178944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
3 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178944 3 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178944 Supplier Novus Biologicals™ Supplier No. NBP178944

For Research applications

Specifications

Antigen ER associated Degradation
Applications Western Blot
Conjugate Unconjugated
Dilution Western Blot
Formulation See individual datasheets.
Immunogen See individual datasheets.
Quantity 3 Vials
Research Discipline Unfolded Protein Response
Test Specificity This ER Associated Degradation Antibodies Sample Pack includes antibodies to Derlin1, OS9 and HRD1.
Includes 1.NB100-2526: HRD1 Antibody
2.NB100-448: Derlin 1 Antibody
3.NB100-519: OS9 Antibody
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.