Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER Stress/ UPR Antibody Pack, R&D Systems™
SDP

Catalog No. NBP252746 Shop All Novus Products
Change view
Click to view available options
Quantity:
8 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252746 8 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252746 Supplier R&D Systems Supplier No. NBP252746

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Antibody Pack

ER Stress / UPR antibody specifically detects ER Stress / UPR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ER Stress / UPR
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene Alias anti-Growth arrest and DNA damage-inducible protein GADD153, ire1 alpha p Ser724, phospho ire1 alpha, phospho-ire1 alpha, p-ire1 alpha
Immunogen See individual datasheets.
Quantity 8 Vials
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cellular Markers, ER Markers, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR)
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.