Learn More
Cambridge Diagnostic Products, Inc.™ Erado-Sol™ Biological and Chemical Stain Remover
Description
- Biological and Chemical Stain Remover
- Fast-Acting
- Readily Biodegradable Surfactants
- Neutral pH 7-8
- No Bleach, Peroxide or Phosphates
Specifications
Specifications
| Product Type | Biological and Chemical Stain Remover |
| Form | Clear Liquid |
| pH | 7 to 8 |
| Content And Storage | Containers which are open must be carefully resealed and kept upright to prevent leakage |
| For Use With (Application) | Removing stains from hands, glassware, lab equipment, stainless steel, plastic, counters, floors, lab coats, clothing and hospital linens |
| Volume (English) | 16 oz. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.