Learn More
Enzo Life Sciences erbB-2 Recombinant monoclonal antibody (4D5-8) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4900200
Encompass_Preferred
Clone: 4D5-8 (Trastuzumab). Isotype: IgG1κ. Immunogen: A431 cells (human epidermoid carcinoma) (over)expressing EGFR. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.