Learn More
ErbB3 (HER-3) Mouse anti-Human , Alexa Fluor 647, Clone: SGP1, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | ErbB3 (HER-3) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SGP1 |
| Conjugate | Alexa Fluor 647 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | ERBB3; ErbB-3; HER3; c-erbB-3; c-erbB3; human epidermal growth factor receptor 3; receptor tyrosine-protein kinase erbB-3 |
| Host Species | Mouse |
| Immunogen | Human cEerbB3 Recombinant Protein |
| Quantity | 50 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.