Learn More
ACROBiosystems Recombinant Protein;Human ErbB3 / Her3 Protein, Strep Tag;HEK293;25UG;ER3,ERBB3,ErbB-3,HER3,HER-3,LCCS2,MDA-BF-1,MGC88033,c-erbB-3,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3
Supplier:  ACROBiosystems ER3H528825UG
Encompass_Preferred
Recombinant Protein;Human ErbB3 / Her3 Protein, Strep Tag;25UG;Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 71.7 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8661
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.