Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4, Rabbit anti-Mouse, Clone: mab10, Millipore Sigma™

Catalog No. p-7162950 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN248425 25 μL
MABN2484100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN248425 Supplier MilliporeSigma Supplier No. MABN248425UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-ErbB4, clone mAB10, Cat. No. MABN2484, is a highly specific rabbit monoclonal antibody that targets Receptor tyrosine-protein kinase ErbB-4 and has been tested for use in ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Receptor tyrosine-protein kinase erbB-4 (UniProt: Q61527; also known as EC: 2.7.10.1, Proto-oncogene-like protein c-ErbB-4) is encoded by the Erbb4 (also known as Mer4) gene (Gene ID: 13869) in murine species. ErbB-4 is a single-pass type I membrane protein of the EGF receptor family that plays an essential role as cell surface receptor for neuregulins (NRG) and EGF family members. ErbB-4 is monomeric in the absence of bound ligand and it homo- or heterodimerizes with another ERBB family member upon ligand binding. ErbB-4 is autophosphorylated on tyrosine residues in response to ligand binding that triggers receptor dimerization and autophosphorylation at specific tyrosine residues, which then serve as binding sites for scaffold proteins and effectors. ErbB-4 is reported to regulate the development of the heart, the central nervous system, and the mammary gland. It is shown to be involved in gene transcription, cell proliferation, differentiation, migration, and apoptosis. It is expressed in the developing and adult cerebral cortex, and represents the major NRG receptor in central neurons and regulates a diverse array of neuronal processes, including migration, differentiation, neurotransmission, and synaptic plasticity. NRG-1 and ErbB-4 are essentials of a signaling pathway that is critical for the proper development of inhibitory cortical and hippocampal circuits. ErbB-4 expression is reported to be restricted to GABAergic neurons in the cerebral cortex where it plays a role in regulating GABAergic synaptic development and interneuron migration. ErbB-4 is synthesized with a signal peptide (aa 1-25), which is cleaved off in the mature form. The mature form has an extracellular domain (aa 26-652), a short transmembrane domain (aa 653-673), and a long cytoplasmic tail (aa 674-1308). Its protein kinase domain is localized in the cytoplasmic tail (aa 718-985). Three isoforms of ErbB-4 have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ErbB4
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mab10
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected ErbB4 in mouse embryo tissue sections.Western Blotting Analysis: A representative lot detected ErbB4 in Western Blotting applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).ELISA Analysis: A representative lot detected ErbB4 in ELISA applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).Immunohistochemistry Analysis: A representative lot detected ErbB4 in Immunohistochemistry applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).Immunoprecipitation Analysis: A representative lot immunoprecipitated ErbB4 in Immunoprecipitation applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).
Formulation Purified rabbit monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Receptor tyrosine-protein kinase erbB-4;Proto-oncogene-like protein c-ErbB-4
Gene Symbols Erbb4;Mer4
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to 204 amino acids from the C-terminal region of murine Receptor tyrosine-protein kinase erbB-4.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_034284.1
Test Specificity Clone mAB10 is a rabbit monoclonal antibody that detects mouse Receptor tyrosine-protein kinase erbB-4. It targets an epitope with in the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.