Learn More
Strem, An Ascensus Company CAS# 10031-51-3. 100g. Erbium(III) nitrate hydrate (99.9%-Er) (REO). MFCD00149691
SDP
Supplier:  Strem, An Ascensus Company 936808100G
Encompass
CAS# 10031-51-3. 100g. Erbium(III) nitrate hydrate (99.9%-Er) (REO). MFCD00149691. Molecular Weight: 353.29. Molecular Formula: Er(NO3)3∙XH2O. Color/form: pink xtl. Strem# 93-6808. http://www.strem.com/catalog/v/93-6808/
Catalog No. 50-901-16167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.