Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium pieces, 99.9% (REO)

Catalog No. aa0011109
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle under Argon
Packaged under argon in glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA0011109 10 g Glass Bottle under Argon
AA0011118 50 g Packaged under argon in glass bottle
AA0011103 1 g Packaged under argon in glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA0011109 Supplier Thermo Scientific Chemicals Supplier No. 00011109
Only null left
Add to Cart

CAS: 7440-52-0 | Er | 167.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Density 9.066 g/cm3
Quantity 10 g
Packaging Glass Bottle under Argon
Form Pieces
Assay Percent Range 99.9% (REO)
Chemical Name or Material Erbium, Rare Earth Oxides Content
Hazard Category H228
Hazard Statement Keep container tightly closed and dry.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-160-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.